missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTER Monoclonal Antibody (OTI1G4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
2851.68 NOK - 6373.49 NOK
Specifications
| Antigen | PTER |
|---|---|
| Clone | OTI1G4 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17507884
|
Origene Technologies
TA813176S |
30 μL |
2851.68 NOK
30µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17587874
|
Origene Technologies
TA813176 |
100 μL |
6373.49 NOK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PTER Monoclonal antibody specifically detects PTER in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| PTER | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| AI790318; HPHRP; Mpr56-1; Parathion hydrolase-related protein; phosphotriesterase related; phosphotriesterase-related protein; Pter; resiniferatoxin-binding phosphotriesterase-related protein; resiniferatoxin-binding, phosphotriesterase-related; resiniferatoxin-binding, phosphotriesterase-related protein; resiniferotoxin-binding phosphotriesterase-related protein; Rpr1; Rpr-1 | |
| Human recombinant protein fragment corresponding to amino acids 1-242 of human PTER (NP_001001484) produced in E.coli. | |
| Primary | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| PTER |
| OTI1G4 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Rat | |
| Q60866, Q63530, Q96BW5 | |
| 19212, 63852, 9317 | |
| IgG1 | |
| Protein A/G | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title