missing translation for 'onlineSavingsMsg'
Learn More

PTER Monoclonal Antibody (OTI1G4), TrueMAB™, OriGene

Product Code. 17507884
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
Unit Size:
100µL
30µL
Product Code. Quantity unitSize
17507884 30 μL 30µL
17587874 100 μL 100µL
2 options
This item is not returnable. View return policy

Product Code. 17507884

Brand: Origene Technologies TA813176S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

PTER Monoclonal antibody specifically detects PTER in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen PTER
Applications Western Blot
Classification Monoclonal
Clone OTI1G4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene PTER
Gene Accession No. Q60866, Q63530, Q96BW5
Gene Alias AI790318; HPHRP; Mpr56-1; Parathion hydrolase-related protein; phosphotriesterase related; phosphotriesterase-related protein; Pter; resiniferatoxin-binding phosphotriesterase-related protein; resiniferatoxin-binding, phosphotriesterase-related; resiniferatoxin-binding, phosphotriesterase-related protein; resiniferotoxin-binding phosphotriesterase-related protein; Rpr1; Rpr-1
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 1-242 of human PTER (NP_001001484) produced in E.coli.
Purification Method Protein A/G
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19212, 63852, 9317
Target Species Human, Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.