missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSMA/GCPII Mouse anti-Human, Rat, CoraLite 488, Clone: 3G4E12, Proteintech
Mouse Monoclonal Antibody
1696.32 NOK - 5307.92 NOK
Specifications
| Antigen | PSMA/GCPII |
|---|---|
| Clone | 3G4E12 |
| Applications | Immunofluorescence |
| Classification | Monoclonal |
| Conjugate | CoraLite 488 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17711299
|
Proteintech
CL488-66678-20UL |
20 μL |
1696.32 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17669866
|
Proteintech
CL488-66678-100UL |
100 μL |
5307.92 NOK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PSMA/GCPII Monoclonal antibody apecifically detetects PSMA/GCPII antigen in Human, Rat Samples. It is validated for Immunofluorescence.Specifications
| PSMA/GCPII | |
| Immunofluorescence | |
| CoraLite 488 | |
| Mouse | |
| PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3. | |
| 2346 | |
| PSMA/GCPII fusion protein Ag16594 | |
| Primary | |
| Store at -20°C. Avoid exposure to light. |
| 3G4E12 | |
| Monoclonal | |
| Liquid | |
| Human, Rat | |
| BC025672 | |
| FOLH1 | |
| IgG1 | |
| Protein G purification |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title