missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSMA/GCPII Mouse anti-Human, Rat, CoraLite 488, Clone: 3G4E12, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-66678-20UL
This item is not returnable.
View return policy
Description
PSMA/GCPII Monoclonal antibody apecifically detetects PSMA/GCPII antigen in Human, Rat Samples. It is validated for Immunofluorescence.Specifications
| PSMA/GCPII | |
| Monoclonal | |
| 1000 ug/mL | |
| PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3. | |
| FOLH1 | |
| PSMA/GCPII fusion protein Ag16594 | |
| 20 μL | |
| 2346 | |
| Store at -20°C. Avoid exposure to light. | |
| IgG1 |
| Immunofluorescence | |
| 3G4E12 | |
| CoraLite 488 | |
| BC025672 | |
| Mouse | |
| Protein G purification | |
| Primary | |
| Human, Rat | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction