missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Prothymosin alpha Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10480-1-AP-20UL
This item is not returnable.
View return policy
Description
Prothymosinalpha(PTalpha) is a nuclear protein that is widely expressed in mammalian tissues, including kidney, liver, spleen, normal lymphocytes, human T cell leukemia virus-infected T cells and myeloma cells. The human PTalpha gene maps to chromosome 2 and encodes a protein that exhibits punctuated nuclear distribution,which correlates to transcription sites. PTalpha is a chromatin-remodeling protein that was initially thought to mediate T lymphocyte maturation,but subsequently has been shown to be involved in cellcycle progression,proliferation and cell differentiation. PTalpha is thought to be transported into the nucleus by the karyopherin beta1-Rch-1 complex, where it associates with Histones H2A, H2B, H3 and H4. Also, PTalpha is phosphorylated on Thr 7 and Thr 12 or 13 by Prothymosin alpha-phosphorylating kinase (PTalphaK) in amitogen activating pathway. The amino terminus of PTalpha iscleaved to produce a secreted, biologically active peptide thymosin alpha1,whichmay be used as an immunomodulatorin cancer patients and patients with chronic active hepatitis,or as an immunoen hancer of vaccinesin immunocompromised individuals.Specifications
| Prothymosin alpha | |
| Polyclonal | |
| Unconjugated | |
| PTMA | |
| Prothymosin alpha, prothymosin, alpha, PTMA, TMSA | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 5757 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.4 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P06454 | |
| PTMA | |
| Prothymosin alpha Fusion Protein Ag0732 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction