missing translation for 'onlineSavingsMsg'
Learn More

Prothymosin alpha Rabbit anti-Human, Polyclonal, Proteintech

Product Code. p-7088014 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16806893 20 μL 20µL
16896883 150 μL 150µL
2 options
This item is not returnable. View return policy

Product Code. 16806893

Brand: Proteintech 104801AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Prothymosinalpha(PTalpha) is a nuclear protein that is widely expressed in mammalian tissues, including kidney, liver, spleen, normal lymphocytes, human T cell leukemia virus-infected T cells and myeloma cells. The human PTalpha gene maps to chromosome 2 and encodes a protein that exhibits punctuated nuclear distribution,which correlates to transcription sites. PTalpha is a chromatin-remodeling protein that was initially thought to mediate T lymphocyte maturation,but subsequently has been shown to be involved in cellcycle progression,proliferation and cell differentiation. PTalpha is thought to be transported into the nucleus by the karyopherin beta1-Rch-1 complex, where it associates with Histones H2A, H2B, H3 and H4. Also, PTalpha is phosphorylated on Thr 7 and Thr 12 or 13 by Prothymosin alpha-phosphorylating kinase (PTalphaK) in amitogen activating pathway. The amino terminus of PTalpha iscleaved to produce a secreted, biologically active peptide thymosin alpha1,whichmay be used as an immunomodulatorin cancer patients and patients with chronic active hepatitis,or as an immunoen hancer of vaccinesin immunocompromised individuals.

Specifications

Antigen Prothymosin alpha
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene PTMA
Gene Accession No. P06454
Gene Alias Prothymosin alpha, prothymosin, alpha, PTMA, TMSA
Gene Symbols PTMA
Host Species Rabbit
Immunogen Prothymosin alpha Fusion Protein Ag0732
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5757
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.