Learn More
Prosulfuron, TRC
Product Code. 17993595
Change view
Click to view available options
Quantity:
50 mg
100 mg
1 g
Unit Size:
100mg
1g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17993595 1 g 1g
17963195 100 mg 100mg
17951815 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17993595 Supplier Toronto Research Chemicals Supplier No. TRCP8386881G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 94125-34-5
Molecular Formula C15 H16 F3 N5 O4 S
Molecular Weight (g/mol) 419.38
Synonym N-[[(4-Methoxy-6-methyl-1,3,5-triazin-2-yl)amino]carbonyl]-2-(3,3,3-trifluoropropyl)benzenesulfonamide, CGA 152005, N-(4-Methoxy-6-methyl-1,3,5-triazin-2-yl)-N'-[2-(3,3,3-trifluoroprop-1-yl)benzenesulfonyl]urea, Peak, Peak 57WG, Pik, Prosulfuron
IUPAC Name 1-(4-methoxy-6-methyl-1,3,5-triazin-2-yl)-3-[2-(3,3,3-trifluoropropyl)phenyl]sulfonylurea
SMILES COc1nc(C)nc(NC(=O)NS(=O)(=O)c2ccccc2CCC(F)(F)F)n1
Specifications
Quantity 1 g
InChI Formula InChI=1S/C15H16F3N5O4S/c1-9-19-12(22-14(20-9)27-2)21-13(24)23-28(25,26)11-6-4-3-5-10(11)7-8-15(16,17)18/h3-6H,7-8H2,1-2H3,(H2,19,20,21,22,23,24)
Formula Weight 419.0875
Chemical Name or Material Prosulfuron