accessibility menu, dialog, popup

UserName

CH3ONCH3NHNOHNSOOFFFN

CAS RN 94125-34-5

IUPAC Name: 3-(4-methoxy-6-methyl-1,3,5-triazin-2-yl)-1-[2-(3,3,3-trifluoropropyl)benzenesulfonyl]urea
Synonyms: prosulfuron
Molecular Weight (g/mol): 419.38
Molecular Formula: C15H16F3N5O4S
InChi Key: LTUNNEGNEKBSEH-UHFFFAOYSA-N
SMILES: COC1=NC(C)=NC(NC(=O)NS(=O)(=O)C2=CC=CC=C2CCC(F)(F)F)=N1
Molecular Weight (g/mol) 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
Concentration 
  • (1)
Element 
  • (1)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results