missing translation for 'onlineSavingsMsg'
Learn More

Prohibitin 2 Mouse anti-Human, Mouse, Rat, Clone: 1D9C7, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66424-1-IG-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16898285

  • 1415.13 NOK / 20µL
Estimated Shipment: 10-10-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

PHB2 acts as a mediator of transcriptional repression by nuclear hormone receptors via recruitment of histone deacetylases. This protein functions as an estrogen receptor (ER)-selective coregulator that potentiates the inhibitory activities of antiestrogens and represses the activity of estrogens. It competes with NCOA1 for modulation of ER transcriptional activity. It is probably involved in regulating mitochondrial respiration activity and in aging.
Specifications

Specifications

Prohibitin 2
Monoclonal
0.7 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O35129, Q5XIH7, Q99623
PHB2
Prohibitin 2 Fusion Protein Ag2977
20 μL
Primary
Human, Rat, Mouse
Antibody
IgG1
Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence
1D9C7
Unconjugated
PHB2
BAP, Bap37, D prohibitin, p22, PHB2, PNAS 141, prohibitin 2, REA
Mouse
Protein G
RUO
11331, 114766, 12034
-20°C
Liquid
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers