missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Prohibitin 2 Mouse anti-Human, Mouse, Rat, Clone: 1D9C7, Proteintech
Mouse Monoclonal Antibody
1710.33 NOK - 4836.10 NOK
Specifications
| Antigen | Prohibitin 2 |
|---|---|
| Clone | 1D9C7 |
| Concentration | 0.7 mg/mL |
| Applications | Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16898285
|
Proteintech
66424-1-IG-20UL |
20 μL |
1710.33 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16888285
|
Proteintech
66424-1-IG-150UL |
150 μL |
4836.10 NOK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PHB2 acts as a mediator of transcriptional repression by nuclear hormone receptors via recruitment of histone deacetylases. This protein functions as an estrogen receptor (ER)-selective coregulator that potentiates the inhibitory activities of antiestrogens and represses the activity of estrogens. It competes with NCOA1 for modulation of ER transcriptional activity. It is probably involved in regulating mitochondrial respiration activity and in aging.Specifications
| Prohibitin 2 | |
| 0.7 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| BAP, Bap37, D prohibitin, p22, PHB2, PNAS 141, prohibitin 2, REA | |
| PHB2 | |
| IgG1 | |
| Protein G | |
| Antibody |
| 1D9C7 | |
| Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Human, Rat, Mouse | |
| O35129, Q5XIH7, Q99623 | |
| 11331, 114766, 12034 | |
| Prohibitin 2 Fusion Protein Ag2977 | |
| Primary | |
| -20°C | |
| PHB2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title