Learn More
BINDER™ Pump Chamber with VAP 5 Vacuum Pump
Pump Chamber with VAP 5 Vacuum Pump - 230 V/50-60 Hz, for mounting vacuum pumps including chemical membrane pump VAP 5 with nominal air flow 3.6 m3;/hr., final pressure 3 mbar, with separator and emission condenser, suitable for flammable solvents and vacuum pump connection kit (ATEX).
Brand:  BINDER™ 8012-2063
147849.37 NOK valid until 2024-04-30
Use promo code "21656" to get your promotional price.
Additional Details : Weight : 69.00000kg
 View more versions of this product
Product Code. 17550916
  • 158790.00 NOK / Each
Estimated Shipment: 14-05-2024
Description
Pump chamber with VAP 5 vacuum pump for use with vacuum pumps with chemical membranes.
  • Nominal air flow
  • With separator and emission condenser
  • Suitable for flammable solvents
  • Includes vacuum pump connection kit (ATEX)
Specifications
Specifications
Pump chamber with vacuum pump VAP 5 - 230 V / 50-60 Hz option model, Pump chamber for mounting vacuum pumps including chemical membrane pump VAP 5 with nominal air flow 3.6 m3/hour, final pressure 3 Mbar, with separator and emission condenser, suitable for flammable solvents and vacuum pump connection kit (ATEX).
50/60 Hz
IP 55
BINDER Chambers VDL 115 Vacuum Drying Chamber
3.6 m3/hr.
Vacuum Pump
3 Mbar
230 V