Learn More
Invitrogen™ E-Gel™ Power Snap Electrophoresis System Starter Kit, DC Safe 1%
Invitrogen™ E-Gel™ Power Snap Electrophoresis System Starter Kit, DC Safe 1%, provides everything you need to quickly start using precast 1% E-Gel™ Double Comb Agarose Gels with SYBR Safe DNA Gel Stain on the system for a streamlined gel electrophoresis and gel imaging workflow.
Brand:  Invitrogen™ G8371ST
24622.80 NOK valid until 2024-06-28
Use promo code "21649" to get your promotional price.
Additional Details : Weight : 3.80000kg
Product Code. 17125989
  • 34680.00 NOK / Each
Estimated Shipment: 20-06-2024
Description
The kit includes:
  • Ten E-Gel™ Double Comb Agarose Gels with SYBR Safe DNA Gel Stain, 1% (Cat. No. A42347)
  • E-Gel™ Power Snap Electrophoresis System (Cat. No. G8300)
  • E-Gel™ 1-Kb Plus Express DNA Ladder with E-Gel™ Sample Loading Buffer (1X) (Cat. No. 10488091)
Specifications
Specifications
• Ten E-Gel Double Comb Agarose Gels with SYBR Safe DNA Gel Stain, 1% (Cat. No. A42347)
• E-Gel Power Snap Electrophoresis System (Cat. No. G8300)
• E-Gel 1-Kb Plus Express DNA Ladder with E-Gel Sample Loading Buffer (1X) (Cat. No. 10488091)
Store at room temperature.
E-Base
Mini
Electrophoresis System Starter Kit