Learn More
RETSCH Zirconium Cover
Brand:  RETSCH 03.107.0320
48687.50 NOK valid until 2024-12-31
Use promo code "23265" to get your promotional price.
Additional Details : Weight : 1.00000kg
Product Code. 10671964
  • 51250.00 NOK / Each
Estimated Shipment: 27-08-2024
Specifications
Cover
For 25 mL Grinding Bowls
Zirconium Oxide