Learn More
RETSCH Stainless Steel Push-fit Feed Chutes
Brand:  RETSCH 03.729.0040
51646.75 NOK valid until 2024-12-31
Use promo code "23265" to get your promotional price.
Additional Details : Weight : 0.20000kg
 View more versions of this product
Product Code. 10074731
  • 54365.00 NOK / Each
Estimated Shipment: 20-08-2024
Specifications
Push-fit Feed Chute
For Holder 75
Stainless Steel
210 x 75/40 mm
210 mm
75/40 mm