Learn More
Invitrogen™ ProcartaPlex™ NHP Control Set, 37plex
Product Code. 17108905
Click to view available options
Quantity:
1 set
Unit Size:
1 set
Product Code. 17108905
Brand: Invitrogen™ EPX37040045CTR
The ProcartaPlex Non-Human Primate Control Set 37plex contains a high and low control for all analytes included in the Non-Human Primate Cytokine/Chemokine/Growth Factor 37-Plex ProcartaPlex Panel.
The ProcartaPlex Non-Human Primate Control Set 37plex contains a high and low control for all analytes included in the Non-Human Primate Cytokine/Chemokine/Growth Factor 37-Plex ProcartaPlex Panel. For a list of analytes included in this set, please refer to the TDS in the Documents section.
Reported application
  • Multiplex Immunoassay
TRUSTED_SUSTAINABILITY
Specifications
Product Type Multiplex Control Set
Quantity 1 set
Species Non-Human Primate
For Use With (Equipment) Luminex™ Instruments
Product Line ProcartaPlex
Content And Storage
• 1 vial NHP Control High A
• 1 vial NHP Control High B
• 1 vial NHP Control Low A
• 1 vial NHP Control Low B
Store at 2°C to 8°C.
Shipping Condition Wet Ice