Learn More
Novus Biologicals™ Primate Universal Protein Whole Tissue Lysate (Adult Whole Normal)
Product Code. 18267641 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
4 Vials
Unit Size:
Pack of 4
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18267641 4 Vials Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18267641 Supplier Novus Biologicals™ Supplier No. NB82089207
  • ONLINE EXCLUSIVE PRICE
    7530.00 NOK
    7121.10 NOK / Pack of 4
    Save 408.90 NOK
    5% Off
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
Add to basket
The Primate Universal Protein Whole Tissue Lysate (Adult Whole Normal) is derived from primate. It has been validated for the following applications: Western Blot, Immunoprecipitation.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Western Blot
Tissue Universal
Quantity 4 Vials
Storage Buffer HEPES (pH 7.9), MgCl2, KCl, EDTA, Sucrose, Glycerol, Sodium deoxycholate, NP-40, and a cocktail of protease inhibitors
Test Specificity Universal Protein Lysate: Monkey Normal Tissues
Cellular Fraction Whole
Tissue Condition Normal
Cross Reactivity Monkey
Host Species Primate
Concentration 5 mg/mL
Product Type Primate Universal Protein Whole Tissue Lysate (Adult Normal)
Content And Storage -80°C. Avoid freeze-thaw cycles.
Life Stage Adult
Protein State Native
Show More Show Less
For Research Use Only