Learn More
JULABO™ PRESTO™ A30 Temperature Control System/Process System
Product Code. 15224363
Change view
Click to view available options
Temperature Range (Metric):
-30°C to +250°C
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Temperature Range (Metric) Unit Size
15224363 -30°C to +250°C Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15224363 Supplier JULABO™ Supplier No. 9420300
Estimated Shipment: 12-01-2027
Julabo™ PRESTO™ A30 Temperature Control System/Process System allows extremely fast compensation of exothermic and endothermic reactions. Lab users benefit from high flow rates, constant pressure, and a controlled build-up of pump pressure.
  • For highly precise, external temperature applications
  • Rapid heating and cooling
  • Wide working temperature ranges without changing fluids
  • Highest performance with small footprint
  • Space-saving design optimizes space utilization in your lab
  • 5,7” industrial color TFT touch screen
  • USB (Host und Device)
  • Ethernet
  • SD-Card slot
  • RS232
  • Modbus
  • RS485 (Accessory)
  • Profibus DP (Accessory)
  • Stand-by input
  • Removable venting grid for simplified removal of dust
  • Permanent internal monitoring and self-lubricating pumps contribute to long service life
Specifications
Certifications/Compliance Classification III (FL) according to DIN 12876-1
Height (Metric) 62 cm
Includes RS232, SD memory card, USB, Ethernet, Modbus
Length (Metric) 59 cm
Temperature (Metric) Maximum 250°C
Temperature Range (Metric) -30°C to +250°C
Width (Metric) 25 cm
Dimensions (L x W x H) 59 x 25 x 62 cm
Model PRESTO™ A30
Weight (Metric) 62 kg
Display Type TFT Touchscreen
Type Temperature Control System
For Use With (Application) Ideal solution for highly precise, external temperature application
Voltage 230 V