Learn More
Julabo™ PRESTO Temperature Control System
Product Code. 15284363
Change view
Click to view available options
Temperature Range (Metric):
-80°C to +250°C
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Temperature Range (Metric) Unit Size
15284363 -80°C to +250°C Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15284363 Supplier Julabo™ Supplier No. 9421801.T
 Restricted Item
Estimated Shipment: 12-01-2027
Highly precise temperature control and rapid temperature change capacity
Julabo PRESTO™ units offer highly precise temperature control and rapid temperature change capacity, making them ideal for applications such as reactor temperature control, substance testing or temperature simulation studies. Highly efficient components permit extremely fast compensation for both exothermic and endothermic reactions. Laboratory users benefit from high flow rates, constant pressure, and a controlled build-up of pump pressure, While permanent internal monitoring and self-lubricating pumps ensure a long service life. The integrated 5.7 inch industrial touch screen displays critical operational information clearly and concisely for extra user-friendly operation.
  • For highly precise, external temperature applications
  • Rapid heating and cooling
  • Wide working temperature ranges without changing fluids
  • Highest performance with small footprint
  • Space-saving design optimizes space utilisation in your lab
  • 5.7 inch industrial color TFT touch screen
  • USB (host and device) and Ethernet connectivity
  • SD-Card slot
  • RS232 / optional RS485 / optional Profibus DP
  • Stand-by input
  • Available voltage versions: 3 x 400V/50Hz, 3 x 230V/50Hz, 3 x 230V/60Hz
Specifications
Certifications/Compliance Classification III (FL) according to DIN 12876-1
Height (Metric) 126 cm
Length (Metric) 65 cm
Temperature (Metric) Maximum 250°C
Temperature Range (Metric) -80°C to +250°C
Width (Metric) 43 cm
Model PRESTO™ W80t
Weight (Metric) 162 kg
Display Type TFT Touchscreen
Type Temperature Control System
For Use With (Application) Ideal solution for highly precise, external temperature application
Voltage 3 x 230/400 V