Learn More
JULABO™ PRESTO™ W80 Temperature Control System/Process System
Product Code. 15274363
Change view
Click to view available options
Temperature Range (Metric):
-80°C to +250°C
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Temperature Range (Metric) Unit Size
15274363 -80°C to +250°C Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15274363 Supplier JULABO™ Supplier No. 9421801
 Restricted Item
Estimated Shipment: 12-01-2027
Made for highly precise temperature control and rapid temperature changes. Allows extremely fast compensation of exothermic and endothermic reactions. Lab users benefit from high flow rates, constant pressure, and a controlled build-up of pump pressure.
  • For highly precise, external temperature applications
  • Rapid heating and cooling
  • Wide working temperature ranges without changing fluids
  • Highest performance with small footprint
  • Space-saving design optimizes space utilization in your lab
  • 5,7” industrial color TFT touch screen
  • USB (Host und Device)
  • Ethernet
  • SD-Card slot
  • RS232
  • Modbus
  • RS485 (Accessory)
  • Profibus DP (Accessory)
  • Stand-by input
  • Water cooled
  • Permanent internal monitoring and self-lubricating pumps contribute to a long service life
Specifications
Certifications/Compliance Classification III (FL) according to DIN 12876-1
Includes RS232, SD memory card, USB, Ethernet, Modbus
Temperature (Metric) Maximum 250°C
Temperature Range (Metric) -80°C to +250°C
Dimensions (L x W x H) 65 x 43 x 126cm
Model PRESTO™ W80
Weight (Metric) 159 kg
Display Type TFT Touchscreen
Type Temperature Control System
For Use With (Application) Ideal solution for highly precise, external temperature application
Voltage 230 V