Specifications
| Capacity (Metric) | 2 mL |
| Particle Size | 0.5mm |
| Volume Range (Metric) | 300 μL to 1.8 mL |
| Material | Glass |
| Dnase Free | Yes |
| For Use With (Application) | Used for processing yeast, cells, fungi, bacteria, spores |
| For Use With (Equipment) | Bead Mill 4 and 24 Homogenizers |
| RNase Free | Yes |
| Product Type | Tough Microorganism Lysing Mix Tubes |