missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRDX1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1710.33 NOK - 4836.10 NOK
Specifications
| Antigen | PRDX1 |
|---|---|
| Concentration | 0.14 mg/mL |
| Applications | Immunofluorescence, Flow Cytometry, Immunoprecipitation, Immunocytochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16855174
|
Proteintech
15816-1-AP-20UL |
20 μL |
1710.33 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16845174
|
Proteintech
15816-1-AP-150UL |
150 μL |
4836.10 NOK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This gene encodes a member of the peroxiredoxin family of antioxidant enzymes, which reduce hydrogen peroxide and alkyl hydroperoxides. The encoded protein may play an antioxidant protective role in cells, and may contribute to the antiviral activity of CD8(+) T-cells. This protein may have a proliferative effect and play a role in cancer development or progression. Three transcript variants encoding the same protein have been identified for this gene.Specifications
| PRDX1 | |
| Immunofluorescence, Flow Cytometry, Immunoprecipitation, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human, Rat, Mouse | |
| P35700, Q06830, Q63716 | |
| 117254, 18477, 5052 | |
| PRDX1 Fusion Protein Ag8602 | |
| Primary | |
| -20°C | |
| PRDX1 |
| 0.14 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| MSP23, NKEF A, NKEFA, PAG, PAGA, PAGB, peroxiredoxin 1, PRDX1, PRX1, PRXI, TDPX2, Thioredoxin peroxidase 2 | |
| Prdx1 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title