missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
This gene encodes a member of the peroxiredoxin family of antioxidant enzymes, which reduce hydrogen peroxide and alkyl hydroperoxides. The encoded protein may play an antioxidant protective role in cells, and may contribute to the antiviral activity of CD8(+) T-cells. This protein may have a proliferative effect and play a role in cancer development or progression. Three transcript variants encoding the same protein have been identified for this gene.
Specifications
Specifications
| Antigen | PRDX1 |
| Applications | Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation |
| Classification | Polyclonal |
| Concentration | 0.14 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | PRDX1 |
| Gene Accession No. | P35700, Q06830, Q63716 |
| Gene Alias | MSP23, NKEF A, NKEFA, PAG, PAGA, PAGB, peroxiredoxin 1, PRDX1, PRX1, PRXI, TDPX2, Thioredoxin peroxidase 2 |
| Gene Symbols | Prdx1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?