Learn More
PRDM5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 14126-1-AP-150UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16882624
  • 4593.76 NOK / 150µL
Estimated Shipment: 22-05-2024
Description
PRDM5, a novel member of PR domain protein family, contains a PR-domain at the-NH2 terminus followed by 16 zinc finger motifs. It is found to have a role in cell differentiation and tumorigenesis. Reports suggest PRDM5 causes G2/M arrest and apoptosis upon infection of tumor cells and has been found to be silenced in breast, ovarian and lung cancers, suggesting a potential clinical significance.
Specifications
Specifications
PRDM5
Polyclonal
Unconjugated
PRDM5
PFM2, PR domain containing 5, PR domain containing protein 5, PRDM5
Rabbit
Antigen Affinity Chromatography
RUO
11107, 689788, 70779
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.35 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9CXE0, Q9NQX1
Prdm5
PRDM5 Fusion Protein Ag5277
150 μL
Primary
Human, Mouse, Rat