missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRDM5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1611.84 NOK - 4593.76 NOK
Specifications
Antigen | PRDM5 |
---|---|
Concentration | 0.35 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16892624
|
Proteintech
14126-1-AP-20UL |
20 μL |
1611.84 NOK
20µL |
Estimated Shipment: 06-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16882624
|
Proteintech
14126-1-AP-150UL |
150 μL |
4593.76 NOK
150µL |
Estimated Shipment: 06-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
PRDM5, a novel member of PR domain protein family, contains a PR-domain at the-NH2 terminus followed by 16 zinc finger motifs. It is found to have a role in cell differentiation and tumorigenesis. Reports suggest PRDM5 causes G2/M arrest and apoptosis upon infection of tumor cells and has been found to be silenced in breast, ovarian and lung cancers, suggesting a potential clinical significance.Specifications
PRDM5 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
Q9CXE0, Q9NQX1 | |
11107, 689788, 70779 | |
PRDM5 Fusion Protein Ag5277 | |
Primary | |
-20°C | |
PRDM5 |
0.35 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
PFM2, PR domain containing 5, PR domain containing protein 5, PRDM5 | |
Prdm5 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |