missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRDM5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14126-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
PRDM5, a novel member of PR domain protein family, contains a PR-domain at the-NH2 terminus followed by 16 zinc finger motifs. It is found to have a role in cell differentiation and tumorigenesis. Reports suggest PRDM5 causes G2/M arrest and apoptosis upon infection of tumor cells and has been found to be silenced in breast, ovarian and lung cancers, suggesting a potential clinical significance.Specifications
PRDM5 | |
Polyclonal | |
Unconjugated | |
PRDM5 | |
PFM2, PR domain containing 5, PR domain containing protein 5, PRDM5 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
11107, 689788, 70779 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.35 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9CXE0, Q9NQX1 | |
Prdm5 | |
PRDM5 Fusion Protein Ag5277 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |