missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PR repeat Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 23979-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
This antibody detects the PRPRPRPRP peptide with dilution 1:4,500 in Elisa.
The C9orf72 "GGGGCC" repeat sequence codes five repeat peptide "GA repeat; GAGAGAGAGA", "GP repeat; GPGPGPGPG", "GR repeat; GRGRGRGRG", "AP repeat; APAPAPAPA" and "PR repeat; PRPRPRPRP". It was described previously that aggregated forms of poly-GA and poly-GP proteins do not enter the separation gel.Specifications
PR repeat | |
Polyclonal | |
Unconjugated | |
C9RAN, GGGCC, poly PR, poly-PR, PR repeat, PRPRPRPRP | |
Peptide | |
20 μL | |
Primary | |
-20°C | |
Liquid |
Western Blot, ELISA, Immunofluorescence, Immunohistochemistry | |
0.25 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
Human | |
Antibody | |
IgG |