missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PR repeat Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1611.84 NOK - 4593.76 NOK
Specifications
Antigen | PR repeat |
---|---|
Concentration | 0.25 mg/mL |
Applications | ELISA, Immunofluorescence, Immunohistochemistry, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16802585
|
Proteintech
23979-1-AP-20UL |
20 μL |
1611.84 NOK
20µL |
Estimated Shipment: 08-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16892575
|
Proteintech
23979-1-AP-150UL |
150 μL |
4593.76 NOK
150µL |
Estimated Shipment: 08-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This antibody detects the PRPRPRPRP peptide with dilution 1:4,500 in Elisa.
The C9orf72 "GGGGCC" repeat sequence codes five repeat peptide "GA repeat; GAGAGAGAGA", "GP repeat; GPGPGPGPG", "GR repeat; GRGRGRGRG", "AP repeat; APAPAPAPA" and "PR repeat; PRPRPRPRP". It was described previously that aggregated forms of poly-GA and poly-GP proteins do not enter the separation gel.Specifications
PR repeat | |
ELISA, Immunofluorescence, Immunohistochemistry, Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
C9RAN, GGGCC, poly PR, poly-PR, PR repeat, PRPRPRPRP | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
0.25 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Peptide | |
Primary | |
-20°C |