missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPP1R14B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18476-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene is thought to play a role in a complex developmental pathway and the regulation of genes involved in cerebellar development. Mutations in this gene have been associated with autosomal recessive spinocerebellar ataxia.Specifications
| PPP1R14B | |
| Polyclonal | |
| Unconjugated | |
| PPP1R14B | |
| PHI 1, PLCB3N, PNG, PPP1R14B, SOM172 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 18938, 259225, 26472 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q62084, Q8K3F3, Q96C90 | |
| Ppp1r14b | |
| PPP1R14B Fusion Protein Ag13373 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction