Learn More
PPP1R14B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 18476-1-AP-20UL
 View more versions of this product
Product Code. 16858504
  • 1710.33 NOK / 20µL
Description
This gene is thought to play a role in a complex developmental pathway and the regulation of genes involved in cerebellar development. Mutations in this gene have been associated with autosomal recessive spinocerebellar ataxia.
Specifications
Specifications
PPP1R14B
Polyclonal
Unconjugated
PPP1R14B
PHI 1, PLCB3N, PNG, PPP1R14B, SOM172
Rabbit
Antigen Affinity Chromatography
RUO
18938, 259225, 26472
-20°C
Liquid
Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q62084, Q8K3F3, Q96C90
Ppp1r14b
PPP1R14B Fusion Protein Ag13373
20 μL
Primary
Mouse, Human, Rat