Learn More
Fisherbrand™ Pour-Boat Weighing Dishes
Product Code. 15708187 Shop All Fisherbrand Products
Change view
Click to view available options
Dimensions (L x W x H):
43 x 58 x 13 mm
108 x 83 x 26 mm
83 x 132 x 26 mm
Unit Size:
Pack of 500
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W x H) unitSize
15708187 83 x 132 x 26 mm Pack of 500
15798177 43 x 58 x 13 mm Pack of 500
15718187 108 x 83 x 26 mm Pack of 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15708187
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Provide accurate pour-outs with minimal sample loss. Pour-Boat Weighing Dishes are polystyrene boats featuring smooth surfaces.
  • Pour-spout design easily dispenses samples without flexing the dish.
  • Flat-bottom dishes are made from 12-mil thick polystyrene for better strength, stability, and handling.
  • Ideal for solid product weighing.
  • Biologically inert and resistant to dilute and weak acids, aqueous solutions, alcohols, and bases.
  • Temperature range -10°C to +70°C (14°F to 158°F).
  • 1 color 3 sizes.
  • Can be used with microwave moisture analyzers.
  • FDA/food-grade compliant.
  • Pack of 500
Specifications
Autoclavable Not Autoclavable
Bottom Shape Flat
Color White
Material Polystyrene
Shape Boat
Sterility Non-sterile
Type Weighing Dish
For Use With (Equipment) Microwave Moisture Analyzer
Quantity 500 Pk.
Temperature Range (English) 14°F to 158°F
Temperature Range (Metric) -10°C to +70°C
Thickness 12 mil
Dimensions (L x W x H) 83 x 132 x 26 mm
Length (English) 3.3 in.
Length (Metric) 83 mm
Width (English) 5.2 in.
Width (Metric) 132 mm
Height (Metric) 26 mm
Height (English) 1 in.