Learn More
Memmert™ Potential-Free Contact
Product Code. 17169934 Shop All Memmert Products
Change view
Click to view available options
For Use With (Equipment):
HPP1060eco
HPP110eco
HPP1400eco
HPP2200eco
HPP260eco
HPP410eco
HPP750eco
ICO105
ICO150
ICO240
ICO50
IPP1060eco
IPP110eco
IPP1400eco
IPP2200eco
IPP260eco
IPP410eco
IPP750eco
Unit Size:
Each
Product Code. For Use With (Equipment) unitSize
17169934 ICO105 Each
17100145 IPP1400eco Each
17100285 IPP750eco Each
17110165 IPP2200eco Each
17119844 HPP410eco Each
17129704 HPP110eco Each
17130005 ICO240 Each
17139754 HPP2200eco Each
17140075 IPP1060eco Each
17150115 IPP110eco Each
17160035 ICO50 Each
17169654 HPP1060eco Each
17169794 HPP260eco Each
17179884 HPP750eco Each
17180195 IPP260eco Each
17180235 IPP410eco Each
17199724 HPP1400eco Each
17199964 ICO150 Each
18 options
Product Code. 17169934
Brand: Memmert™ ICO105H6
Only null left
Add to basket
Add to basket
For combination error message (e.g. supply failure,sensor fault, fuse)
  • Accessory
  • For combination error message (e.g. supply failure,sensor fault, fuse)
Specifications
Description Combination error message
For Use With (Equipment) ICO105