Learn More
Fisherbrand™ Portable Balance
Product Code. 15909355 Shop All Fisherbrand Products
Change view
Click to view available options
Capacity:
220 g
620 g
2200 g
5200 g
Model:
FCS2200
FCS221
FCS5200
FCS621
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity Model unitSize
15909355 2200 g FCS2200 Each
15939345 220 g FCS221 Each
15949345 620 g FCS621 Each
15919355 5200 g FCS5200 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 15909355
Only null left
Add to basket
Add to basket
Portable Balance with a slim, stackable design provides basic weighing functionality and competitive performance.
  • Intuitive two-button design makes for easy operation
  • Slim, space-saving design allows for stackable storage
  • Enlarged weighing platform can adequately handle large samples and a variety of objects
TRUSTED_SUSTAINABILITY
Tekniske data
Capacity 2200 g
Display Type LCD
Readability 1.000000 g
For Use With (Application) Weighing
Includes 3 AA (LR6) Batteries
Model FCS2200
Stabilization Time 2.000 sec.
Temperature Range (English) 64°F to 77°F
Temperature Range (Metric) 18°C to 25°C
Warranty 2 Years
Weighing Modes Gram, Pound, Ounce, Ounce Troy, Pennyweight
Dimensions (D x W x H) 205 x 140 x 41 mm (8.1 x 5.5 x 1.6 in.)
Width (English) Pan 5 in.
Width (Metric) Pan 125 mm
Depth (English) Pan 5.2 in.
Depth (Metric) Pan 132 mm
Weight (English) 0.9 lb.
Weight (Metric) 0.4 kg
Vis mere Vis mindre
Produkttitel
Vælg et problem
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.