Learn More
Thermo Scientific™ POROS™ Caprylate Mixed-Mode Cation Exchange Chromatography Resin
Shop All Thermo Scientific ProductsDescription
POROS Caprylate Mixed-Mode Chromatography Resin is designed to remove moderate to high levels of aggregates during downstream processing of therapeutic antibodies in flow-through mode. As a mixed-mode, weak cation-exchange resin, POROS Caprylate resin effectively removes other impurities, such as host cell proteins and a wide range of high molecular weight species.
Features of POROS Caprylate Mixed-Mode resin include:
- Selective capture of aggregates and hydrophobic high-molecular-weight impurities
- Flow-through design that helps save buffer and material costs
- Excellent scalability and high productivity
- Wide operational range
- Non-animal-derived
POROS Caprylate Mixed-Mode resin operates in flow-through mode which reduces costs, especially when working with more engineered monoclonal antibody derivatives. The unique selectivity of the caprylate ligand contributes to a high yield of the targeted monomer while using established cation exchange resin buffer systems.
Main characteristics
- Matrix: cross-linked poly(styrene-divinylbenzene) POROS beads
- Average particle size: 50 μm
- Ligand: octanoic acid
- Binding capacity: >20 g lysozyme/L resin
- Typical buffers: typical cation exchange buffers, pH 4.5–7.0, conductivity 10–30 mS/cm
- Clean-in-place: 0.5 M NaOH
- Formulation buffer: 0.1 M NaOH
- Mechanical resistance: 100 bar (10 MPa, 1450 psi)
POROS Caprylate Mixed-Mode resin is manufactured using a synthetic manufacturing process that is free of animal components.
Specifications
Specifications
| Particle Size | 50 μm |
| Product Line | POROS |
| Type | Bulk Resin |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.