Learn More
Fisherbrand™ Porcelain Plate For Desiccators
Product Code. 15584645 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Diameter (Metric) Outer:
90 mm
140 mm
190 mm
235 mm
280 mm
For Use With (Equipment):
Desiccator (Dia.: 100mm)
Desiccator (Dia.: 150mm)
Desiccator (Dia.: 200mm)
Desiccator (Dia.: 250mm)
Desiccator (Dia.: 300mm)
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Outer For Use With (Equipment) Unit Size
15584645 190 mm Desiccator (Dia.: 200mm) Each
15594645 235 mm Desiccator (Dia.: 250mm) Each
15574645 140 mm Desiccator (Dia.: 150mm) Each
15564645 90 mm Desiccator (Dia.: 100mm) Each
15504655 280 mm Desiccator (Dia.: 300mm) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 15584645
Estimated Shipment: 14-12-2026
Add to basket
Add to basket
For use with desiccators
  • For use with desiccators
Specifications
Diameter (Metric) Outer 190 mm
For Use With (Equipment) Desiccator (Dia.: 200mm)
Product Type Desiccator Plate