Learn More
Applied Biosystems™ POP-7™ Polymer for Protein Quality Analysis
Shop All Applied Biosystems ProductsDescription
Includes
Radio frequency identification (RFID) label.
- Easy to use: Pre-formulated and in a single-use pouch, which not only saves time but ensures reliability and consistency
- Robust formulation: Formulated for biomolecular analysis applications
- Replenishable: Allows capillaries to be used multiple times
- Includes radio frequency identification (RFID) label: Easily track important information such as part/lot number, samples remaining, and expiry dates (displayed on software dashboard)
Specifications
Specifications
| Content And Storage | Store at 2–8°C. |
| Description | POP-7™ Polymer For 3500xL Genetic Analyzer |
| For Use With (Equipment) | 3500xL Genetic Analyzer for Protein Quality Analysis |
| For Use With (Application) | Glycan Analysis, Protein Quality Analysis |
| No. of Samples | 960 |
| Product Line | POP-7 |
| Product Type | Polymer |
| Quantity | 960 Samples |
| Shipping Condition | Wet Ice |
For research use only