Learn More
Fisherbrand™ Polyurethane Foam Stoppers
Product Code. p-8013160 Shop All Fisherbrand Products
Change view
Click to view available options
Diameter (Metric):
15 mm
25 mm
30 mm
35 mm
40 mm
55 mm
70 mm
Unit Size:
Pack of 100
Product Code. Diameter (Metric) unitSize
12944311 15 mm Pack of 100
12954311 25 mm Pack of 100
12964291 30 mm Pack of 100
12984291 40 mm Pack of 100
12974291 35 mm Pack of 100
12904301 55 mm Pack of 100
12914301 70 mm Pack of 100
7 options
Product Code. 12944311
Only null left
Add to basket
Add to basket
Formed by polymerization in the presence of a foaming agent.
  • Made up of polyurethane
  • Rigid and flexible
Specifications
Diameter (Metric) 15 mm
Height (Metric) 30 mm
Quantity 100/Pk.
Material Polyurethane
Product Type Plug Stopper