Learn More
Fisherbrand™ Polyurethane Foam Stoppers
Product Code. 12944311 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Diameter (Metric):
15 mm
25 mm
30 mm
35 mm
40 mm
55 mm
70 mm
Unit Size:
Pack of 100
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Unit Size
12944311 15 mm Pack of 100
12954311 25 mm Pack of 100
12964291 30 mm Pack of 100
12984291 40 mm Pack of 100
12974291 35 mm Pack of 100
12904301 55 mm Pack of 100
12914301 70 mm Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 12944311
  • ONLINE EXCLUSIVE PRICE
    86.10 NOK
    69.10 NOK / Pack of 100
    Save 17.00 NOK
    19% Off
Estimated Shipment: 09-11-2026
Add to basket
Add to basket
Formed by polymerization in the presence of a foaming agent.
  • Made up of polyurethane
  • Rigid and flexible
Specifications
Diameter (Metric) 15 mm
Height (Metric) 30 mm
Quantity 100/Pk.
Material Polyurethane
Product Type Plug Stopper