Learn More
Fisherbrand™ Polypropylene Yellow Pipet Tip E
Product Code. 11587442 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Volume (Metric):
1 to 200 μL
Unit Size:
Pack of 1000
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
11587442 1 to 200 μL Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11587442
Estimated Shipment: 15-12-2026
Add to basket
Add to basket
Make pipetting simple and safer for even the most sensitive applications. Fisherbrand™ Polypropylene Yellow Pipet Tips E are perfectly fit for manual or electronic, single or multichannel pipets.
  • Made of high quality polypropylene
  • Tight fit due to the precisely fitting cone
  • Optimal handling due to the stable shape
  • Autoclavable
  • Guaranteed dust-free due to special ionization treatment
  • Available pre-sterilized or non-sterile
  • Excellent transparency due to high quality surfaces
  • Minimal residual wetting due to controlled quality of raw materials
  • Excellent volume accuracy due to precise tip orifice
  • Practice-oriented tip volumes
  • Aeroject™ tips with special filters made of hydrophobic PE-HD as
  • Aerosol barrier and for protection against contamination
  • Suitable for all commonly used pipet types
Specifications
Volume (Metric) 1 to 200 μL
Color Yellow