Learn More
Eppendorf™ Polypropylene V-Bottom 384 Well Microplate
GREENER_CHOICE
Product Code. 10678404 Shop All Eppendorf Products
Click to view available options
Color:
Gray, White Bottom, White Bottom
White, Black Bottom, Black Bottom
White, Clear Bottom
Certified:
PCR clean
Sterile
Quantity:
240/Pk.
80/Pk.
Unit Size:
Pack of 240
Pack of 80
Product Code. 10678404
missing translation for 'mfr': Eppendorf™ 0030625307
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Certified PCR clean
Format Standard
Array 16 x 24
Autoclavable Autoclavable
Color White, Clear Bottom
Material Polypropylene
No. of Wells 384
Well Shape Conical Bottom (V-well)
Quantity 240/Pk.