Learn More
Eppendorf™ Polypropylene V-Bottom 384 Well Microplate
Shop All Eppendorf Products
Click to view available options
Color:
Gray, White Bottom, White Bottom
White, Black Bottom, Black Bottom
White, Clear Bottom
Certified:
PCR clean
Sterile
Quantity:
240/Pk.
80/Pk.
Unit Size:
Pack of 240
Pack of 80
Specifications
Specifications
| Certified | PCR clean |
| Format | Standard |
| Array | 16 x 24 |
| Autoclavable | Autoclavable |
| Color | Gray, White Bottom, White Bottom |
| Material | Polypropylene |
| No. of Wells | 384 |
| Well Shape | Conical Bottom (V-well) |
| Quantity | 80/Pk. |