Learn More
Fisherbrand™ Polypropylene Storage Boxes with lid, Autoclavable, for EPA-Vials
Product Code. 12652465 Shop All Fisherbrand Products
Change view
Click to view available options
Dimensions (L x W x H):
130 x 130 x 80 mm
130 x 130 x 105 mm
For Use With (Equipment):
For 20ml EPA vials
For 30ml, 40ml EPA vials
No. of Places:
10 cavities
16 cavities
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W x H) For Use With (Equipment) No. of Places unitSize
12652465 130 x 130 x 105 mm For 30ml, 40ml EPA vials 10 cavities Each
12652505 130 x 130 x 80 mm For 20ml EPA vials 16 cavities Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12652465
Temperature resistant from -80° up to +100°.
  • Storage Box
  • Includes cover
  • Feature alphanumeric indexing for easier vial identification
  • Stackable for efficient storage
TRUSTED_SUSTAINABILITY
Specifications
Color Violet
For Use With (Equipment) For 30ml, 40ml EPA vials
No. of Places 10 cavities
Dimensions (L x W x H) 130 x 130 x 105 mm