Learn More
Fisherbrand™ Polypropylene Pipet Finntip™
Product Code. 11567462 Shop All Fisherbrand Products
Change view
Click to view available options
Volume (Metric):
1000 to 5000 μL
2000 to 10,000 μL
Unit Size:
Pack of 100
Pack of 250
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
11567462 2000 to 10,000 μL Pack of 100
11527462 1000 to 5000 μL Pack of 250
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11567462
Spotlight Offers
Use promo code 28036
View all promotions
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
Make pipetting simple and safer for even the most sensitive applications. Fisherbrand™ Polypropylene Pipet Finntip™s are perfectly fit for manual or electronic, single or multichannel pipets.
  • Made of high quality polypropylene
  • Tight fit due to the precisely fitting cone
  • Optimal handling due to the stable shape
  • Autoclavable
  • Guaranteed dust-free due to special ionization treatment
  • Excellent transparency due to high quality surfaces
  • Minimal residual wetting due to controlled quality of raw materials
  • Excellent volume accuracy due to precise tip orifice
  • Practice-oriented tip volumes
  • Aeroject™ tips with special filters made of hydrophobic PE-HD as
  • Aerosol barrier and for protection against contamination
  • Suitable for all commonly used pipet types
Specifications
Volume (Metric) 2000 to 10,000 μL
Sterility Non-sterile
Color Natural
Autoclavable Autoclavable
Product Line FinnTip