Learn More
Eppendorf™ Polypropylene F-Bottom 384 Well Microplate
6710.00 NOK - 17430.00 NOK
Specifications
Format | Standard |
---|---|
Array | 16 x 24 |
Autoclavable | Autoclavable |
Color | White, Clear Bottom |
Material | Polypropylene |
Product Code | Brand | Sterility | Color | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Sterility | Color | Price | Quantity & Availability | ||||
10065283
|
Eppendorf™
0030621107 |
Non-sterile | White, Clear Bottom |
6710.00 NOK
Pack of 80 |
Estimated Shipment: 05-06-2024
Log in to see stock. |
||||
10723814
|
Eppendorf™
0030622103 |
Sterile | White, Clear Bottom |
7750.00 NOK
Pack of 80 |
Estimated Shipment: 05-06-2024
Log in to see stock. |
||||
10095043
|
Eppendorf™
0030626109 |
Sterile | White, Clear Bottom |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
10380554
|
Eppendorf™
0030625102 |
Non-sterile | White, Clear Bottom |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Specifications
Standard | |
Autoclavable | |
Polypropylene | |
Flat Bottom (F-well) |
16 x 24 | |
White, Clear Bottom | |
384 |