Learn More
Eppendorf™ Polypropylene F-Bottom 384 Well Microplate
Shop All Eppendorf Products
Click to view available options
Color:
White, Clear Bottom
Sterility:
Non-sterile
Sterile
Quantity:
240/Pk.
80/Pk.
Unit Size:
Pack of 240
Pack of 80
Specifications
Specifications
| Format | Standard |
| Array | 16 x 24 |
| Autoclavable | Autoclavable |
| Color | White, Clear Bottom |
| Material | Polypropylene |
| No. of Wells | 384 |
| Sterility | Non-sterile |
| Well Shape | Flat Bottom (F-well) |
| Quantity | 80/Pk. |