Learn More
Eppendorf™ Polypropylene Adaptor
Product Code. 10206531 Shop All Eppendorf Products
Change view
Click to view available options
For Use With (Equipment):
Bottle Thread 32 to 28 mm
Bottle thread 25 mm
Bottle thread GL 32 to GL 27
Bottle thread GL 32 to GL 45
Bottle thread GL 45 to GL 35
Bottle thread GL 45 to GL 38
Bottle thread GL 45 to S 40
Bottleneck 19/26
Thread GL 32 to NS 24/29
Thread GL 32 to NS 29/32
Unit Size:
Each
Product Code. For Use With (Equipment) unitSize
10206531 Bottle thread GL 32 to GL 27 Each
10023211 Bottle thread 25 mm Each
10738052 Bottle Thread 32 to 28 mm Each
10586911 Bottle thread GL 32 to GL 45 Each
10277011 Thread GL 32 to NS 24/29 Each
10217111 Thread GL 32 to NS 29/32 Each
10237111 Bottle thread GL 45 to GL 38 Each
10691312 Bottle thread GL 45 to GL 35 Each
10328661 Bottle thread GL 45 to S 40 Each
10338611 Bottleneck 19/26 Each
10 options
Product Code. 10206531
Brand: Eppendorf™ 4960800139
Only null left
Specifications
Product Type Adapter
Material Polypropylene
For Use With (Equipment) Bottle thread GL 32 to GL 27
Quantity 1 Ea.