Learn More
Fisherbrand™ Delrin™ Full-Size Test Tube Racks, 72 x 13 mm
GREENER_CHOICE
Product Code. 15360380 Shop All Thermo Scientific CO₂ Incubators Products
Change view
Click to view available options
Color:
Blue
Green
Magenta
White
Yellow
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
15360380 Green Each
15380380 Yellow Each
15340380 White Each
15350380 Blue Each
15370380 Magenta Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 15360380
Molded in a single, continuous piece. Delrin™ Full-Size Test Tube Racks, 72 x 13 mm are highly resistant to heat and chemical attack.
  • Delrin™ fiberglass-reinforced polyoxymethylene (POM)
  • Offers the highest chemical and heat resistance (-40 to +140°C)
  • Unique tear drop shaped openings for better retention of tubes
  • Autoclavable
  • Molded coordinates for tube orientation
  • Stable design for use in water baths
  • Flat labeling area for easier identification
Specifications
Autoclavable Autoclavable
Color Green
Holds Tubes (13 mm), Tubes (5 to 10 mL)
Material Fiberglass-reinforced POM
Positions 72
Shape Teardrop-shaped Openings
Stackable Yes, When Empty
Sterility Non-sterile
For Use With (Equipment) 5 to 10 mL Test Tubes
No. per Case 1/Cs.
Quantity 8 Pk.
Dimensions (L x W x H) 104 x 202 x 59 mm
Length (Metric) 104 mm
Length (English) 4.1 in.
Width (English) 8 in.
Height (English) 2.3 in.
Diameter (Metric) Opening 13 mm
Product Type Test Tube Rack
Shelves None
Height (Metric) 59 mm
Array 6 x 12
Width (Metric) 202 mm