Learn More
Macherey-Nagel™ Polyethylene 50° shore A Hardness Snap Ring Caps
Brand:  Macherey-Nagel™ 702718.1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 12450430
  • 357.00 NOK / Pack of 100
Estimated Shipment: 27-05-2024
Specifications
Blue
Yes
Snap Ring Cap
1 mm
N11
Polyethylene
Septum
100 Pk.
Snap Ring Cap