Learn More
Macherey-Nagel™ Polyethylene 50° shore A Hardness Snap Ring Caps
357.00 NOK
Specifications
Material | Polyethylene |
---|---|
Septum | Yes |
Includes | Septum |
Insert Type | Snap Ring Cap |
Quantity | 100 Pk. |
Product Code | Brand | Color | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Price | Quantity & Availability | |||||
12450430
|
Macherey-Nagel™
702718.1 |
Blue |
357.00 NOK
Pack of 100 |
Estimated Shipment: 15-05-2024
Log in to see stock. |
|||||
12440430
|
Macherey-Nagel™
702718 |
Transparent |
357.00 NOK
Pack of 100 |
Estimated Shipment: 08-05-2024
Log in to see stock. |
|||||
Specifications
Polyethylene | |
Septum | |
100 Pk. | |
Snap Ring Cap |
Yes | |
Snap Ring Cap | |
1 mm | |
N11 |