Learn More
Macherey-Nagel™ Polyethylene 50° shore A Hardness Snap Ring Caps
Brand: Macherey-Nagel™ 702718
Additional Details : Weight : 0.01000kg
Specifications
Transparent | |
Yes | |
Snap Ring Cap | |
1 mm | |
N11 |
Polyethylene | |
Septum | |
100 Pk. | |
Snap Ring Cap |