Learn More
HUBER Polycarbonate Bath
Product Code. 10744826
Change view
Click to view available options
Model:
106 A
108 A
110 A
112 A
118 A
130 A
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Model Unit Size
10744826 118 A Each
10112214 106 A Each
10131085 108 A Each
10744636 110 A Each
10666137 112 A Each
10374245 130 A Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 10744826 Supplier HUBER Supplier No. 30526
  • ONLINE EXCLUSIVE PRICE
    8535.00 NOK
    7419.80 NOK / Pack of 1
    Save 1115.20 NOK
    13% Off
Estimated Shipment: 29-12-2026
A range of polycarbonate baths in various capacities
Polycarbonate bath
Specifications
Description Bath
Material Polycarbonate
Model 118 A